missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Dibenzoyl-L-tartaric Acid Monohydrate 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 160250050

 View more versions of this product

Catalog No. AC160250050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(-)-Dibenzoyl-L-tartaric acid monohydrate
62708-56-9,2743-38-6
100.0
-113° to -117° (20°C, 589nm) (c=5, ethanol)
−115° (20°C c=5,ethanol)
≥97.5% (on dry substance)
98%
C18H14O8·H2O
376.33
98%
97.5
Authentic
4 to 5.5%
Powder
White
5g
[C6H5CO2CH(CO2H)-]2·H2O
MFCD00149119

Chemical Identifiers

62708-56-9,2743-38-6
100.0
C18H14O8·H2O
376.33
(-)-Dibenzoyl-L-tartaric acid monohydrate, 98%
97.5
98%
[C6H5CO2CH(CO2H)-]2·H2O
MFCD00149119
≥97.5% (on dry substance)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA