missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Dibenzoyl-L-tartaric acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438120250

 View more versions of this product

Catalog No. AC438120250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(-)-Dibenzoyl-L-tartaric acid
Glass bottle
1% Max.
150.0°C to 151.6°C
2743-38-6
100.0
C18H14O8
YONLFQNRGZXBBF-ZIAGYGMSSA-N
(2R,3R)-2,3-dibenzoyloxybutanedioic acid
75969
Solid or Crystalline Powder
Authentic
− 116.70
Brown to White
25g
97.5
97.5% min. (HPLC)
--dibenzoyl-l-tartaric acid, 2,3-bis benzoyloxy tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, dibenzoyltartaric acid, l-dibenzoyl tartaric acid, o,o-dibenzoyl-+-tartaric acid, unii-3q14b666vm
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O
358.31
358.31
98%

Chemical Identifiers

2743-38-6
358.31
--dibenzoyl-l-tartaric acid, 2,3-bis benzoyloxy tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, dibenzoyltartaric acid, l-dibenzoyl tartaric acid, o,o-dibenzoyl-+-tartaric acid, unii-3q14b666vm
(2R,3R)-2,3-dibenzoyloxybutanedioic acid
C18H14O8
YONLFQNRGZXBBF-ZIAGYGMSSA-N
75969
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 220-374-

SDS Documents