missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibenzothiophene, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 112541000
Specifications
Dibenzothiophene | |
Authentic | |
332°C to 333°C | |
97°C to 100°C | |
132-65-0 | |
C12H8S | |
17,72 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,benzene,chloroform | |
C1=CC=C2C(=C1)C3=CC=CC=C3S2 | |
184.26 | |
CHEBI:23681 | |
Crystalline Powder and/or Chunks |
170°C | |
Plastic bottle | |
Gray-Green to White | |
100g | |
97.5% min. (GC) | |
MFCD00004969 | |
.alpha.-thiafluorene, 1,1'-biphenyl-2,2'-diyl sulfide, 2,2'-biphenylylene sulfide, 9-thiafluorene, alpha-thiafluorene, dibenzo b,d thiophene, diphenylene sulfide, unii-z3d4aj1r48, usan | |
IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
dibenzothiophene | |
3023 | |
184.26 | |
98% |
Chemical Identifiers
132-65-0 | |
184.26 | |
IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
3023 | |
dibenzothiophene |
C12H8S | |
MFCD00004969 | |
.alpha.-thiafluorene, 1,1'-biphenyl-2,2'-diyl sulfide, 2,2'-biphenylylene sulfide, 9-thiafluorene, alpha-thiafluorene, dibenzo b,d thiophene, diphenylene sulfide, unii-z3d4aj1r48, usan | |
CHEBI:23681 | |
C1=CC=C2C(=C1)C3=CC=CC=C3S2 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
Collect spillage.
Dispose of contents/
GHS Signal Word: Warning
EINECSNumber : 205-072-9