Learn More
Dibenzothiophene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1228822
Description
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Dibenzothiophene | |
97°C to 100°C | |
170°C (338°F) | |
MFCD00004969 | |
UN3077 | |
diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene | |
IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
dibenzothiophene | |
3023 | |
184.25 |
132-65-0 | |
332°C to 333°C | |
C12H8S | |
100 g | |
121101 | |
Soluble in water. | |
C1=CC=C2C(=C1)C3=CC=CC=C3S2 | |
184.256 | |
CHEBI:23681 | |
98% |
Chemical Identifiers
132-65-0 | |
184.256 | |
IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
3023 | |
dibenzothiophene |
C12H8S | |
MFCD00004969 | |
diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene | |
CHEBI:23681 | |
C1=CC=C2C(=C1)C3=CC=CC=C3S2 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 205-072-9
RTECSNumber : HQ3490550
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only