Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzothiophene-4-carboxaldehyde, 98%

Chemical Identifiers

Chemical Name or Material Dibenzothiophene-4-carboxaldehyde
CAS 23985-81-1
Formula Weight 212.27
MDL Number MFCD00961967
Beilstein 1567946
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6451503

  SDS
alfa aesar™
H6451503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6451506

  SDS
alfa aesar™
H6451506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64515MD

  SDS
alfa aesar™
H64515MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.27
Chemical Name or Material Dibenzothiophene-4-carboxaldehyde
Physical Form Powder
Assay 98%
Color Pale yellow
Quantity 1g
Sensitivity Air Sensitive
MDL Number MFCD00961967
CAS 23985-81-1
Beilstein 1567946
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.