missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzothiophene-2-boronic acid, 97%

$43.18 - $562.88

Chemical Identifiers

CAS 668983-97-9
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.072
MDL Number MFCD01318982
InChI Key CSLSCVHILGCSTE-UHFFFAOYSA-N
Synonym dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
PubChem CID 2794660
IUPAC Name dibenzothiophen-2-ylboronic acid
SMILES B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64816MD View Documents Alfa Aesar™
H64816MD
250mg Each for $43.18
 
AAH6481603 View Documents Alfa Aesar™
H6481603
1g Each for $128.77
 
AAH6481606 View Documents Alfa Aesar™
H6481606
5g Each for $562.88
 
Specifications

Chemical Identifiers

668983-97-9
228.072
CSLSCVHILGCSTE-UHFFFAOYSA-N
2794660
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
C12H9BO2S
MFCD01318982
dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
dibenzothiophen-2-ylboronic acid

Specifications

White
668983-97-9
MFCD01318982
dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl, boronic acid, b-2-dibenzothienyl, dibenzo b,d thiophen-2-ylboronic acid contains varying amounts of anhydride, dibenzo b,d thien-2-ylboronic acid saltdata: free, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-4-ylboronic acid, ksc352o5l
B(C1=CC2=C(C=C1)SC3=CC=CC=C32)(O)O
228.072
228.07
97%
250mg
C12H9BO2S
15124
CSLSCVHILGCSTE-UHFFFAOYSA-N
dibenzothiophen-2-ylboronic acid
2794660
Powder
Dibenzothiophene-2-boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C