Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzosuberenone, 97%

$34.80 - $213.96

Chemical Identifiers

Chemical Name or Material Dibenzosuberenone
CAS 2222-33-5
Molecular Formula C15H10O
Formula Weight 206.24
MDL Number MFCD00003588
Beilstein 743171
Synonym 5H-Dibenzo[a,d]cyclohepten-5-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2372706

  SDS
alfa aesar™
B2372706
5g Each for $34.80

AAB2372722

  SDS
alfa aesar™
B2372722
100g Each for $213.96
Description & Specifications

Specifications

Formula Weight 206.24
Molecular Formula C15H10O
Chemical Name or Material Dibenzosuberenone
Quantity 5g
Beilstein 743171
RTECS Number HP1147500
CAS 2222-33-5
Assay 97%
Melting Point 88° to 89°C
MDL Number MFCD00003588
Synonym 5H-Dibenzo[a,d]cyclohepten-5-one
TSCA TSCA

Applications
Dibenzosuberenone can be used in the synthesis of a series of potent antidepressants.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.