Learn More
Dibenzofuran, 98%, Thermo Scientific Chemicals
$148.56 - $339.27
Chemical Identifiers
CAS | 132-64-9 |
---|---|
Molecular Formula | C12H8O |
Molecular Weight (g/mol) | 168.195 |
MDL Number | MFCD00004968 |
InChI Key | TXCDCPKCNAJMEE-UHFFFAOYSA-N |
Synonym | dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide |
PubChem CID | 568 |
ChEBI | CHEBI:28145 |
IUPAC Name | dibenzofuran |
SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3O2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1652114
|
Thermo Scientific Chemicals
A1652114 |
25 g |
Each for $148.56
|
|
|||||
AAA1652122
|
Thermo Scientific Chemicals
A1652122 |
100 g |
Each for $339.27
|
|
|||||
Description
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsApplications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.
Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
132-64-9 | |
168.195 | |
TXCDCPKCNAJMEE-UHFFFAOYSA-N | |
568 | |
dibenzofuran |
C12H8O | |
MFCD00004968 | |
dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide | |
CHEBI:28145 | |
C1=CC=C2C(=C1)C3=CC=CC=C3O2 |
Specifications
132-64-9 | |
1.3 | |
130°C (266°F) | |
MFCD00004968 | |
UN3077 | |
dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide | |
TXCDCPKCNAJMEE-UHFFFAOYSA-N | |
dibenzofuran | |
568 | |
168.2 | |
Dibenzofuran |
78°C to 83°C | |
287°C to 289°C | |
C12H8O | |
25 g | |
121100 | |
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents. | |
C1=CC=C2C(=C1)C3=CC=CC=C3O2 | |
168.195 | |
CHEBI:28145 | |
98% |
Safety and Handling
H500
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 205-071-3
RTECSNumber : HP4430000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only