missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibenzofuran-4-boronic acid, 98+%, Thermo Scientific Chemicals
$46.75 - $143.73
Chemical Identifiers
CAS | 100124-06-9 |
---|---|
Molecular Formula | C12H9BO3 |
Molecular Weight (g/mol) | 212.011 |
MDL Number | MFCD00092336 |
InChI Key | ZXHUJRZYLRVVNP-UHFFFAOYSA-N |
Synonym | dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid |
PubChem CID | 2734328 |
IUPAC Name | dibenzofuran-4-ylboronic acid |
SMILES | B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1983003
|
Thermo Scientific Chemicals
L1983003 |
1 g |
Each for $46.75
|
|
|||||
AAL1983006
|
Thermo Scientific Chemicals
L1983006 |
5 g |
Each for $143.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100124-06-9 | |
212.011 | |
ZXHUJRZYLRVVNP-UHFFFAOYSA-N | |
2734328 | |
B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O |
C12H9BO3 | |
MFCD00092336 | |
dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid | |
dibenzofuran-4-ylboronic acid |
Specifications
100124-06-9 | |
C12H9BO3 | |
1 g | |
dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid | |
B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O | |
212.011 | |
212.01 | |
Dibenzofuran-4-boronic acid |
286°C to 291°C | |
MFCD00092336 | |
1645426 | |
ZXHUJRZYLRVVNP-UHFFFAOYSA-N | |
dibenzofuran-4-ylboronic acid | |
2734328 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only