Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzofuran-2-sulfonic acid hydrate, 97%

$61.64 - $61.64

Chemical Identifiers

Chemical Name or Material Dibenzofuran-2-sulfonic acid hydrate
CAS 83863-63-2
Molecular Formula C12H8O4S·xH2O
Formula Weight 248.24 (Anhydrous)
MDL Number MFCD00043577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4389903

  SDS
alfa aesar™
4389903
1g Each for $61.64
Description & Specifications

Specifications

Formula Weight 248.24 (Anhydrous)
Chemical Name or Material Dibenzofuran-2-sulfonic acid hydrate
Melting Point 144° to 145°C
Physical Form Powder
Assay 97%
Quantity 1g
Molecular Formula C12H8O4S·xH2O
Sensitivity Hygroscopic
MDL Number MFCD00043577
CAS 83863-63-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.