missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzofuran-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6490503

 View more versions of this product

Catalog No. AAH6490503



Description

Description

Used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an pharmaceutical intermediate.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

Dibenzofuran-2-carboxaldehyde
74°C
C13H8O2
1 g
Air Sensitive
OVJMIWIVPWPZMN-UHFFFAOYSA-N
dibenzofuran-2-carbaldehyde
220843
98%
5397-82-0
White
MFCD00136272
141044
dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
196.21
196.2
Powder

Chemical Identifiers

5397-82-0
196.21
OVJMIWIVPWPZMN-UHFFFAOYSA-N
220843
C13H8O2
MFCD00136272
dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only