Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzo[b,e]thiepin-11(6H)-one, 98%

$49.11 - $532.59

Chemical Identifiers

Chemical Name or Material Dibenzo[b,e]thiepin-11(6H)-one
CAS 1531-77-7
Molecular Formula C14H10OS
Formula Weight 226.29
MDL Number MFCD02093738
Synonym 11-Oxo-6,11-dihydrodibenzo[b,e]thiepin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2340206

  SDS
alfa aesar™
B2340206
5g Each for $49.11

AAB2340214

  SDS
alfa aesar™
B2340214
25g Each for $158.54

AAB2340222

  SDS
alfa aesar™
B2340222
100g Each for $532.59
Description & Specifications

Specifications

Formula Weight 226.29
Chemical Name or Material Dibenzo[b,e]thiepin-11(6H)-one
Melting Point 86° to 88°C
Boiling Point 175° to 180°C (1mmHg)
Assay 98%
Quantity 5g
Molecular Formula C14H10OS
MDL Number MFCD02093738
CAS 1531-77-7
Synonym 11-Oxo-6,11-dihydrodibenzo[b,e]thiepin
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.