Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzenechromium, C 69.2%, H 5.8%

Chemical Identifiers

Chemical Name or Material Dibenzenechromium
CAS 1271-54-1
Linear Formula (C6H6)2Cr
Formula Weight 208.22
MDL Number MFCD00015485
UN Number UN3391
Synonym Chromium dibenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2413902

  SDS
alfa aesar™
2413902
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2413904

  SDS
alfa aesar™
2413904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2413909

  SDS
alfa aesar™
2413909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.22
UN Number UN3391
Concentration C 69.2%, H 5.8%
Color Black
MDL Number MFCD00015485
Health Hazard 3 GHS P Statement
P210-P222-P280-P370+P378a-P422a-P420
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store contents under inert gas.
Store away from other materials.
Chemical Name or Material Dibenzenechromium
Health Hazard 2 GHS H Statement
H250-H252
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Sensitivity Air & Moisture Sensitive
Synonym Chromium dibenzene
Quantity 0.5g
CAS 1271-54-1
RTECS Number GB5850000
Melting Point 260° to 265°C
Health Hazard 1 Danger
Linear Formula (C6H6)2Cr
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
Physical Form Crystalline
Safety and Handling

Danger

GHS P Statement
P210-P222-P280-P370+P378a-P422a-P420
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store contents under inert gas.
Store away from other materials.

GHS H Statement
H250-H252
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.