missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzenechromium, C 69.2%, H 5.8%

Manufacturer:  Alfa Aesar™ 2413909

 View more versions of this product

Catalog No. AA2413909



Specifications

Specifications

Dibenzenechromium
Black
10g
C12H12Cr
MFCD00015485
bis benzene chromium, bis benzene chromium 0, chromium, bis benzene-8ci, dibenzolchrom, bis benzene chromium 0, bis benzene chromium, chromium, bis h6-benzene-9ci
HVURSIGIEONDKB-UHFFFAOYSA-N
benzene;chromium
10932683
Crystalline
Air & Moisture Sensitive
260°C to 265°C
1271-54-1
(C6H6)2Cr
UN3391
C 69.2%, H 5.8%
C1=CC=CC=C1.C1=CC=CC=C1.[Cr]
208.224
208.22

Chemical Identifiers

1271-54-1
208.224
HVURSIGIEONDKB-UHFFFAOYSA-N
10932683
C1=CC=CC=C1.C1=CC=CC=C1.[Cr]
C12H12Cr
MFCD00015485
bis benzene chromium, bis benzene chromium 0, chromium, bis benzene-8ci, dibenzolchrom, bis benzene chromium 0, bis benzene chromium, chromium, bis h6-benzene-9ci
benzene;chromium
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.

GHS P Statement
P210-P222-P280-P370+P378a-P422a-P420
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store contents under inert gas.
Store away from other materials.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC

RTECSNumber : GB5850000

SDS Documents