missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diazald, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc S1170

Catalog No. S1170



Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diazald80-11-51000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Diazald
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
0.1,99.9
C8H10N2O3S
FFKZOUIEAHOBHW-UHFFFAOYSA-N
N,4-dimethyl-N'-oxobenzene-1-sulfonohydrazide
6628
Methanol-P&T
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
80-11-5,67-56-1
0.1,99.9
MFCD00002050
CN(N=O)S(=O)(=O)C1=CC=C(C)C=C1
214.24
2mL amber prescored glass ampule
1mL

Chemical Identifiers

80-11-5
214.24
FFKZOUIEAHOBHW-UHFFFAOYSA-N
N,4-dimethyl-N'-oxobenzene-1-sulfonohydrazide
C8H10N2O3S
MFCD00002050
6628
CN(N=O)S(=O)(=O)C1=CC=C(C)C=C1
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents
Product Certifications

Product intended for laboratory use only