missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diaza-18-crown-6, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC275821000

 View more versions of this product

Catalog No. AC275821000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Chelation/Complexation reagent
Specifications

Specifications

Diaza-18-crown-6
Glass bottle
111°C to 115°C
98%
97.5% min. (GC)
MFCD00005112
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,chloroform,acetone
C1COCCOCCNCCOCCOCCN1
262.34
262.34
98%
Authentic
White
100mg
23978-55-4
C12H26N2O4
1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane, 7,16-diaza-18-crown-6, cryptand 2.2, cryptand 22, diaza-18-crown-6, kryptofix 22, unii-xly51t1rsz, xly51t1rsz
NLMDJJTUQPXZFG-UHFFFAOYSA-N
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
72805
Shiny Crystalline Powder

Chemical Identifiers

23978-55-4
262.34
NLMDJJTUQPXZFG-UHFFFAOYSA-N
72805
C1COCCOCCNCCOCCOCCN1
C12H26N2O4
MFCD00005112
1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane, 7,16-diaza-18-crown-6, cryptand 2.2, cryptand 22, diaza-18-crown-6, kryptofix 22, unii-xly51t1rsz, xly51t1rsz
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 245-965-0

RTECSNumber : HM1650000

SDS Documents