Learn More
Diaza-18-crown-6, 98%, Thermo Scientific Chemicals
$162.91 - $162.91
Chemical Identifiers
CAS | 23978-55-4 |
---|---|
Molecular Formula | C12H28N2O4 |
Molecular Weight (g/mol) | 264.37 |
MDL Number | MFCD00005112 |
InChI Key | NLMDJJTUQPXZFG-UHFFFAOYSA-P |
Synonym | kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane |
PubChem CID | 72805 |
IUPAC Name | 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane |
SMILES | C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC275825000
|
Thermo Scientific Chemicals
275825000 |
500 mg | Glass bottle |
Each for $162.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
23978-55-4 | |
264.37 | |
NLMDJJTUQPXZFG-UHFFFAOYSA-P | |
72805 | |
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 |
C12H28N2O4 | |
MFCD00005112 | |
kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane | |
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane |
Specifications
23978-55-4 | |
White | |
97.5% min. (GC) | |
C12H28N2O4 | |
500 mg | |
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,chloroform,acetone | |
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 | |
264.37 | |
262.34 | |
Shiny Crystalline Powder |
111°C to 115°C | |
Authentic | |
Glass bottle | |
MFCD00005112 | |
kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane | |
NLMDJJTUQPXZFG-UHFFFAOYSA-P | |
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane | |
72805 | |
98% | |
Diaza-18-crown-6, 98% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 245-965-0
RTECSNumber : HM1650000
RUO – Research Use Only