missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diatrizoic acid, Thermo Scientific Chemicals

A high-osmolality radiocontrast agent

Supplier:  Thermo Scientific Chemicals J6344422

 View more versions of this product

Catalog No. AAJ6344422



Description

Description

Diatrizoic acid is a gradient reagent used for blood cell separation and diagnostic aid (radiopaque medium). Diatrizoic acid may be used as an alternative to barium sulfate for medical imaging of the gastrointestinal tract, such as upper gastrointestinal series and small bowel series.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from oxidizing agents.
Specifications

Specifications

Diatrizoic acid
White
MFCD00069960
14,2992
Soluble in Methanol.
CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I
613.92
CHEBI:53691
Powder
117-96-4
C11H9I3N2O4
100 g
diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate
YVPYQUNUQOZFHG-UHFFFAOYSA-N
3,5-diacetamido-2,4,6-triiodobenzoic acid
2140
613.9

Chemical Identifiers

117-96-4
613.92
YVPYQUNUQOZFHG-UHFFFAOYSA-N
2140
3,5-diacetamido-2,4,6-triiodobenzoic acid
C11H9I3N2O4
MFCD00069960
diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate
CHEBI:53691
CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I
Safety and Handling

Safety and Handling

EINECSNumber : 204-223-6

RTECSNumber : DG5950000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only