missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diatonic Scale Tuning Forks

Harmonious chords familiar to piano players!

Manufacturer:  Fisher Science Education 700100ED

 View more versions of this product

Catalog No. S42015M



Description

Description

The diatonic scale is based on the C- major I chord familiar to piano players and consists of the notes C, E, and G, which sound particularly harmonious when vibrating together. The diatonic scale adjusts these notes so that their frequency ratios are exactly 4:5:6, the frequency ratio that produces the most harmonious C-major chord.

  • Nontarnishing forks are made from lightweight tempered aluminum alloy
  • Each fork is stamped with scale letter and frequency
  • All forks are tuned to within ±0.5% of the frequency stamped on the fork when operating at a temperature of 22°C

Order Info

Based on C=256Hz. Fork length: 27cm.

Specifications

Specifications

100Hz
71°F
Tuning Fork
Aluminum Alloy
22°C
SDS
Documents
Product Certifications