Diatonic Scale Mounted Tuning Forks
Manufacturer:  Fisher Scientific 700256MR
 View more versions of this product
Catalog No. S42051M
Description
Description
Forks vibrate molecules in box cavity!
When tuning forks are mounted on a wooden cavity, the top of the cavity vibrates causing vibrations of air molecules inside of the cavity. These tuning forks are mounted to cavities that resonate at the same frequency as that of the tuning fork.
Note: for simple resonance experiments only, not to be used in pairs.
Specifications
Specifications
C Scale
256Hz
Tuning Fork
Documents