Please login to your online account to display your discounted pricing
Diatonic Scale Mounted Tuning Forks
$91.05 - $91.05
Specifications
Product Type Tuning Fork
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Hertz Price Quantity    
S42051M
fisher science education
700-256MR
C Scale 256Hz Each for $91.05
S42055
fisher science education
SCSM3757
G Scale 384Hz Each for $91.05
S42053M
fisher science education
700-320MR
E Scale 320Hz Each for $91.05
Description & Specifications
Specifications
Product Type Tuning Fork
Forks vibrate molecules in box cavity!
When tuning forks are mounted on a wooden cavity, the top of the cavity vibrates causing vibrations of air molecules inside of the cavity. These tuning forks are mounted to cavities that resonate at the same frequency as that of the tuning fork.
Note: for simple resonance experiments only, not to be used in pairs.