Please login to your online account to display your discounted pricing

Diatomaceous Earth, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$329.75 - $329.75

Chemical Identifiers

Chemical Name or Material Diatomaceous Earth
CAS 68855-54-9
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002717

 
avantor performance
337101
Wide Mouth Amber Glass Bottle 500g Each for $329.75
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Diatomaceous Earth
CAS 68855-54-9
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.30kg
Purity Grade BAKER ANALYZED™ Reagent
Particle Size 48 Micron

    Suitable for use in Pesticide Residue Analysis after Extraction with Petroleum Ether