Please login to your online account to display your discounted pricing

Diatomaceous Earth, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  337101

 View more versions of this product

Catalog No. 02002717

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Diatomaceous Earth
Packaging Wide Mouth Amber Glass Bottle
Purity Grade BAKER ANALYZED™ Reagent
CAS 68855-54-9
Specific Gravity 1L = 2.30kg
Particle Size 48 Micron

Specifications

Chemical Name or Material Diatomaceous Earth
CAS 68855-54-9
Purity Grade BAKER ANALYZED™ Reagent

    Suitable for use in Pesticide Residue Analysis after Extraction with Petroleum Ether