missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diamond powder, synthetic, <1 micron, 99.9% (metals basis), Thermo Scientific Chemicals
Diamond powder is generally used as an abrasive to grind and polish hard surfaces
Supplier: Thermo Scientific Chemicals 01340106
Description
Diamond powder is generally used as an abrasive to grind and polish hard surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diamond powder | |
Powder | |
(metals basis) | |
99.9% |
Synthetic | |
5 g | |
MFCD00211867 |
RUO – Research Use Only