Please login to your online account to display your discounted pricing

Alfa Aesar™ Diamminetetrachlorotitanate(IV), 99%

Chemical Identifiers

Chemical Name or Material Diamminetetrachlorotitanate(IV)
CAS 15719-81-0
Formula Weight 223.75
MDL Number MFCD00145368
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3959304

  SDS
alfa aesar™
3959304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3959309

  SDS
alfa aesar™
3959309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.75
Chemical Name or Material Diamminetetrachlorotitanate(IV)
Physical Form Powder
Assay 99%
MDL Number MFCD00145368
CAS 15719-81-0
UN Number UN3260
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Sensitivity Moisture Sensitive
Boiling Point 120°C (0.1mmHg) (subl.)
Melting Point 240°C (dec.)
Quantity 2g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.