Please login to your online account to display your discounted pricing

Alfa Aesar™ Diamminepalladium(II) nitrite solution, Pd 8-10% w/w (cont. Pd)

Chemical Identifiers

Chemical Name or Material Diamminepalladium(II) nitrite solution
CAS 28068-05-5
Formula Weight 232.47g/mol (Anhydrous)
MDL Number MFCD00038712
UN Number UN2672
Synonym Diamminedinitritopalladate(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11038NJ

 
alfa aesar™
11038-NJ
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Ammonia-like
Quantity ≈0.5g
Color Yellow
Quantity 0.5g
Concentration Pd 8 to 10% w/w (cont. Pd)
Chemical Name or Material Diamminepalladium(II) nitrite solution
CAS 28068-05-5
Physical Form Liquid
Formula Weight 232.47g/mol (Anhydrous)
MDL Number MFCD00038712
UN Number UN2672
Synonym Diamminedinitritopalladate(II)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
TSCA TSCA