Learn More
Diaminomaleonitrile, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2432814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diaminomaleonitrile | |
178°C to 182°C (decomposition) | |
C4H4N4 | |
25 g | |
878333 | |
diaminomaleonitrile, 2,3-diaminomaleonitrile, hydrogen cyanide tetramer, maleonitrile, diamino, ccris 921, 2-butenedinitrile, 2,3-diamino-, 2z, 2z-diaminobut-2-enedinitrile, 2,3-diamino maleonitrile, 2-butenedinitrile, 2,3-diamino-, z, z-2,3-diaminobut-2-enedinitrile | |
C(#N)C(=C(C#N)N)N | |
108.104 | |
108.1 |
1187-42-4 | |
1.36 | |
MFCD00001870 | |
UN3439 | |
Light sensitive | |
DPZSNGJNFHWQDC-ARJAWSKDSA-N | |
(Z)-2,3-diaminobut-2-enedinitrile | |
2723951 | |
98% |
Chemical Identifiers
1187-42-4 | |
108.104 | |
DPZSNGJNFHWQDC-ARJAWSKDSA-N | |
2723951 | |
C(#N)C(=C(C#N)N)N |
C4H4N4 | |
MFCD00001870 | |
diaminomaleonitrile, 2,3-diaminomaleonitrile, hydrogen cyanide tetramer, maleonitrile, diamino, ccris 921, 2-butenedinitrile, 2,3-diamino-, 2z, 2z-diaminobut-2-enedinitrile, 2,3-diamino maleonitrile, 2-butenedinitrile, 2,3-diamino-, z, z-2,3-diaminobut-2-enedinitrile | |
(Z)-2,3-diaminobut-2-enedinitrile |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P312-P330-P363-P501c
H301-H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 214-697-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only