Please login to your online account to display your discounted pricing

Alfa Aesar™ Diallylamine, 98%

$44.45 - $44.45

Chemical Identifiers

Chemical Name or Material Diallylamine
CAS 124-02-7
Molecular Formula (CH2=CHCH2)2NH
Formula Weight 97.16
MDL Number MFCD00008642
UN Number UN2359
Beilstein 773718
Merck Index 14,2971
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA18950AE

  SDS
alfa aesar™
A18950AE*
100mL Each for $44.45

AAA18950AP

  SDS
alfa aesar™
A18950AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 97.16
Molecular Formula (CH2=CHCH2)2NH
CAS 124-02-7
Merck Index 14,2971
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point −88°C
Boiling Point 111° to 112°C
Assay 98%
UN Number UN2359
TSCA TSCA
RTECS Number UC6650000
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H311-H330-H314-H318-H302
Highly flammable liquid and vapour.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Flash Point 7°C (44°F)
Beilstein 773718
Quantity 100mL
MDL Number MFCD00008642
Chemical Name or Material Diallylamine
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIALLYLAMINE

Applications
Diallylamine is used as an intermediate for ion exchange resins, pharmaceuticals, water soluble polymers, rubber chemicals, polymerization initiators and cross-linking agents.

Solubility
Soluble in ethanol and ether. Moderately soluble in water.

Notes
Air Sensitive. store away from water/moisture and air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H330-H314-H318-H302
Highly flammable liquid and vapour.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.