Learn More
Diallylamine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 112042500
Specifications
Diallylamine | |
7°C | |
Glass bottle | |
0.789 | |
Colorless to Yellow | |
250mL | |
124-02-7 | |
C6H11N | |
MFCD00008642 | |
15, 2974 | |
Solubility in water: 90g/L in water. Other solubilities: soluble in diethyl ether, ethanol | |
C=CCNCC=C | |
97.16 | |
97.16 | |
97% |
0.7890g/mL | |
Authentic | |
1.4395 to 1.4415 | |
110.0°C to 112.0°C | |
-88.0°C | |
97% | |
97% | |
(H2C=CHCH2)2NH | |
04, 208 | |
2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, ccris 4776, di-2-propenylamine, diallylamine, n,n-diallylamine, n-2-propenyl-2-propen-1-amine, n-allylprop-2-en-1-amine, unii-n18exb6v6p | |
DYUWTXWIYMHBQS-UHFFFAOYSA-N | |
N-prop-2-enylprop-2-en-1-amine | |
31279 | |
Liquid |
Chemical Identifiers
124-02-7 | |
97.16 | |
DYUWTXWIYMHBQS-UHFFFAOYSA-N | |
31279 | |
C=CCNCC=C |
C6H11N | |
MFCD00008642 | |
2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, ccris 4776, di-2-propenylamine, diallylamine, n,n-diallylamine, n-2-propenyl-2-propen-1-amine, n-allylprop-2-en-1-amine, unii-n18exb6v6p | |
N-prop-2-enylprop-2-en-1-amine |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 204-671-2
RTECSNumber : UC6650000
TSCA : TSCA