Please login to your online account to display your discounted pricing

Diallylamine 97%, ACROS Organics™


Quantity 5mL

Manufacturer: thermo fisher scientific  112040050

 View more versions of this product

Catalog No. AC112040050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 97.16
Physical Form Liquid
Quantity 5mL
Color Clear colorless to yellow
EINECS Number 204-671-2
TSCA TSCA
RTECS Number UC6650000
Density 0.789g/mL
Beilstein 04, 208
Merck Index 15, 2974
Infrared Spectrum Authentic
Synonym Di-2-propenylamine
Refractive Index 1.4395 to 1.4415 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Diallylamine
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H11N
Linear Formula (H2C=CHCH2)2NH
MDL Number MFCD00008642
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Highly flammable liquid and vapor.
CAS 124-02-7

Specifications

Chemical Name or Material Diallylamine
Name Note 97%
CAS 124-02-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11N
Linear Formula (H2C=CHCH2)2NH
Formula Weight 97.16
MDL Number MFCD00008642
Synonym Di-2-propenylamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents