Learn More
Diallyl sulfide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1097122
Description
Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in alcohol, chloroform, ether, and carbon tetrachloride. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Diallyl sulfide | |
-83°C | |
137°C to 138°C | |
Unpleasant | |
1.4895 | |
MFCD00008658 | |
UN1993 | |
14,297 | |
Soluble in alcohol,chloroform,ether,and carbon tetrachloride. Insoluble in water. | |
C=CCSCC=C | |
114.21 | |
CHEBI:4489 | |
98% |
592-88-1 | |
0.889 | |
23°C (73°F) | |
C6H10S | |
CH2=CHCH2SCH2CH=CH2 | |
100 g | |
1736016 | |
diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide | |
UBJVUCKUDDKUJF-UHFFFAOYSA-N | |
3-(prop-2-en-1-ylsulfanyl)prop-1-ene | |
11617 | |
114.21 |
Chemical Identifiers
592-88-1 | |
114.21 | |
UBJVUCKUDDKUJF-UHFFFAOYSA-N | |
11617 | |
C=CCSCC=C |
C6H10S | |
MFCD00008658 | |
diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide | |
CHEBI:4489 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 209-775-1
RTECSNumber : BC4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only