Please login to your online account to display your discounted pricing

Diallyl phthalate, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  276620050

 View more versions of this product

Catalog No. AC276620050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 246.26
Quantity 5mL
Color Clear colorless to light yellow
Beilstein 09, 4120
Density 1.1200g/mL
UN Number 3082
Chemical Name or Material Diallyl phthalate
Molecular Formula C14H14O4
Health Hazard 1 Warning
MDL Number MFCD00008646
Packaging Glass bottle
Assay Percent Range 98%
Infrared Spectrum Authentic
CAS 131-17-9
Synonym 1, 2-Benzenedicarboxylic acid di-2-propenylester
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Specifications

Chemical Name or Material Diallyl phthalate
CAS 131-17-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H14O4
Formula Weight 246.26
MDL Number MFCD00008646
Synonym 1, 2-Benzenedicarboxylic acid di-2-propenylester
Accessories

Warning

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

SDS & Safety Documents