Learn More
Diallyl phthalate, 97%, Thermo Scientific Chemicals
$34.78 - $90.52
Chemical Identifiers
CAS | 131-17-9 |
---|---|
Molecular Formula | C14H14O4 |
Molecular Weight (g/mol) | 246.262 |
MDL Number | MFCD00008646 |
InChI Key | QUDWYFHPNIMBFC-UHFFFAOYSA-N |
Synonym | diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st |
PubChem CID | 8560 |
IUPAC Name | bis(prop-2-enyl) benzene-1,2-dicarboxylate |
SMILES | C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2464822
|
Thermo Scientific Chemicals
B2464822 |
100 g |
N/A
|
|
|||||
AAB2464836
|
Thermo Scientific Chemicals
B2464836 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131-17-9 | |
246.262 | |
QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
8560 | |
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C |
C14H14O4 | |
MFCD00008646 | |
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
bis(prop-2-enyl) benzene-1,2-dicarboxylate |
Specifications
131-17-9 | |
1.12 | |
166°C (330°F) | |
C14H14O4 | |
MFCD00008646 | |
UN3082 | |
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st | |
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C | |
246.262 | |
246.27 | |
Diallyl phthalate |
-70°C | |
160°C to 163°C (4 mmHg) | |
Pungent, Makes Eyes Water | |
1.519 | |
100 g | |
1880877 | |
QUDWYFHPNIMBFC-UHFFFAOYSA-N | |
bis(prop-2-enyl) benzene-1,2-dicarboxylate | |
8560 | |
97% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P311-P330-P333+P313-P363-P501c
H302-H317-H331-H340-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 205-016-3
RTECSNumber : CZ4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only