missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 999-21-3 |
---|---|
Molecular Formula | C10H12O4 |
Molecular Weight (g/mol) | 196.202 |
MDL Number | MFCD00010763 |
InChI Key | ZPOLOEWJWXZUSP-WAYWQWQTSA-N |
Synonym | diallyl fumarate, fumaric acid, diallyl ester, unii-k852lk4c5p, 2-butenedioic acid e-, di-2-propenyl ester, 2-butenedioic acid 2e-, di-2-propenyl ester, butenedioic acid diallyl ester, 1,4-bis prop-2-en-1-yl 2e-but-2-enedioate, 2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester, diallyl malleate, 1,4-bis prop-2-en-1-yl but-2-enedioate |
PubChem CID | 5354266 |
IUPAC Name | bis(prop-2-enyl) (Z)-but-2-enedioate |
SMILES | C=CCOC(=O)C=CC(=O)OCC=C |
Chemical Identifiers
999-21-3 | |
196.202 | |
ZPOLOEWJWXZUSP-WAYWQWQTSA-N | |
5354266 | |
C=CCOC(=O)C=CC(=O)OCC=C |
C10H12O4 | |
MFCD00010763 | |
diallyl fumarate, fumaric acid, diallyl ester, unii-k852lk4c5p, 2-butenedioic acid e-, di-2-propenyl ester, 2-butenedioic acid 2e-, di-2-propenyl ester, butenedioic acid diallyl ester, 1,4-bis prop-2-en-1-yl 2e-but-2-enedioate, 2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester, diallyl malleate, 1,4-bis prop-2-en-1-yl but-2-enedioate | |
bis(prop-2-enyl) (Z)-but-2-enedioate |
Specifications
154°C | |
-47°C | |
999-21-3 | |
MFCD00010763 | |
diallyl fumarate, fumaric acid, diallyl ester, unii-k852lk4c5p, 2-butenedioic acid e-, di-2-propenyl ester, 2-butenedioic acid 2e-, di-2-propenyl ester, butenedioic acid diallyl ester, 1,4-bis prop-2-en-1-yl 2e-but-2-enedioate, 2-butenedioic acid 2e-, 1,4-di-2-propen-1-yl ester, diallyl malleate, 1,4-bis prop-2-en-1-yl but-2-enedioate | |
C=CCOC(=O)C=CC(=O)OCC=C | |
196.202 | |
196.20 | |
≥97.0% (GC) |
Yellow | |
500g | |
C10H12O4 | |
2810 | |
ZPOLOEWJWXZUSP-WAYWQWQTSA-N | |
bis(prop-2-enyl) (Z)-but-2-enedioate | |
5354266 | |
Clear Liquid at 20°C | |
Diallyl Maleate |
Safety and Handling
EINECSNumber : (2)-1110
RTECSNumber : ON0700000
TSCA : Yes