Please login to your online account to display your discounted pricing

Alfa Aesar™ Diallyl maleate, 95%

Chemical Identifiers

Chemical Name or Material Diallyl maleate
CAS 999-21-3
Molecular Formula C10H12O4
Formula Weight 196.21
MDL Number MFCD00010763
UN Number UN2810
Beilstein 1725954
Synonym Maleic acid diallyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL038670B

 
alfa aesar™
L038670B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0386718

 
alfa aesar™
L0386718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0386730

 
alfa aesar™
L0386730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.21
TSCA TSCA
Assay 95%
Molecular Formula C10H12O4
MDL Number MFCD00010763
CAS 999-21-3
UN Number UN2810
Health Hazard 3 GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number ON0700000
Chemical Name or Material Diallyl maleate
Beilstein 1725954
Synonym Maleic acid diallyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 109° to 110°C (3mmHg)
Melting Point −47°C
Flash Point 130°C (266°F)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 1000g

Applications
Diallyl maleate has potential applications in polyester resins, adhesives, and ion exchange resins. When used at low levels, Diallyl Maleate is an effective agent for the promotion of branching in emulsion polymers.

Solubility
Soluble in water 151.2 mg/L @ 25°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H312-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.