Please login to your online account to display your discounted pricing

Alfa Aesar™ Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide


Quantity 5g

Manufacturer: alfa aesar™  L1110606

 View more versions of this product

Catalog No. AAL1110606

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 146.27
Chemical Name or Material Diallyl disulfide
Assay 80%
Molecular Formula C6H10S2
CAS 2179-57-9
Beilstein 1699241
Purity Grade Technical Grade
Assay Percent Notes remainder mainly diallyl sulfide and diallyl trisulfide
Linear Formula CH2=CHCH2S-SCH2CH=CH2
UN Number UN1993
Synonym Allyl disulfide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 180° to 190°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number BB1000000
TSCA TSCA
MDL Number MFCD00008656
Odor Garlic-like
Flash Point 51°C (123°F)
Recommended Storage 2° to 8°C
Quantity 5g

Specifications

Chemical Name or Material Diallyl disulfide
CAS 2179-57-9
Purity Grade Technical Grade
Molecular Formula C6H10S2
Linear Formula CH2=CHCH2S-SCH2CH=CH2
Formula Weight 146.27
MDL Number MFCD00008656
UN Number UN1993
Beilstein 1699241
Synonym Allyl disulfide
Accessories

Warning

GHS P Statement
P260-P280h-P305+P351+P338
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C

SDS & Safety Documents