Diaion™ UBK555(Ca), ion exchange fractionation resin, 8% x-linked, strongly acidic gel type, 2.0meq/mL on PS-DVB, Thermo Scientific™
Supplier:  Thermo Scientific Chemicals 46731A1
Catalog No. AA46731A1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Chromatographic sugar fractionation , fructose/glucose separations, sugar alcohol separations
Specifications
Specifications
Diaion™ UBK555 (Ca), ion exchange fractionation resin
Odorless
Brown
8% cross-linked, strongly acidic gel type, 2.0meq/mL on PS-DVB
220μm
1kg