Diaion™ CRB05(OH), chelating resin, highly porous type, 0.95 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 046721A1
Catalog No. AA46721A1
Description
Description
Boron, arsenic, gallium, and selenium removal
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
300-850μm Beads
Highly porous type, 0.95meq/mL on poly(styrene-divinylbenzene)
300 to 850μm
1 kg
Diaion™ CRB05 (OH), chelating resin
71799-50-3
1.1
Chemical Identifiers
71799-50-3
Safety and Handling
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures