Alfa Aesar™ Diaion™ CR20, chelating resin, highly porous type, 0.4 mmole/ml on poly(styrene-divinylbenzene)
Transitional metal removal without alkali earth metal adsorption
$176.51 - $176.51
Chemical Identifiers
Chemical Name or Material Diaion™ CR20, chelating resin, Highly porous type, 0.4mmole/mL on poly(styrene-divinylbenzene)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA46781A1 SDS Alfa Aesar™
46781A1
1kg Each for $176.51
Specifications Description & Specifications
Chemical Identifiers
Diaion™ CR20, chelating resin, Highly porous type, 0.4mmole/mL on poly(styrene-divinylbenzene)
Specifications
Odorless
Beads
1kg
300 to 1180μm
Pale yellow
Diaion™ CR20, chelating resin, Highly porous type, 0.4mmole/mL on poly(styrene-divinylbenzene)
Safety and Handling
TSCA : TSCA