Learn More
Diacetone-D-glucose, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1556822
Description
It is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is mainly used in biochemical reaction and used as medicine intermediate. It is used in synthesize for below products: L-gulose;1,2:5,6-Di-O-isopropylidene-a-D-gulofuranose; Alllose; ;1,2:5,6-Double-O-Isopropylidene-3-C-(1-Methoxycarbonyl)Ethide-alfa-D-Furanose) ;1,2:5,6-Di-O-isopropylidene-a-D-ribo-hexofuranose-3-ulose;6-deoxy idose.
Solubility
Solubility in hot water gives very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Diacetone-D-glucose | |
107°C to 110°C | |
MFCD00005544 | |
84386 | |
1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol | |
−11° (c=5 in Ethanol) | |
(3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol | |
40469256 | |
≥98% |
582-52-5 | |
C12H20O6 | |
100 g | |
14,2965 | |
KEJGAYKWRDILTF-MQIGXGKASA-N | |
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C | |
260.286 | |
260.29 |
Chemical Identifiers
582-52-5 | |
260.286 | |
KEJGAYKWRDILTF-MQIGXGKASA-N | |
40469256 | |
CC1(OCC(O1)C2C(C3C(O2)OC(O3)(C)C)O)C |
C12H20O6 | |
MFCD00005544 | |
1,2:5,6-diisopropylidene-d-glucose, 1,2:5,6-di-o-isopropylidene-alpha-d-glucofuranose, 3ar,5r,6s,6as-5-4s-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 2,3-d 1,3 dioxol-6-ol | |
(3aR,5S,6S,6aS)-5-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]-2,2-dimethyl-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol |
Safety and Handling
EINECSNumber : 209-486-0
RTECSNumber : LZ4958000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only