Please login to your online account to display your discounted pricing

Diacetone acrylamide, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diacetone acrylamide
Name Note 99%
CAS 2873-97-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H15NO2
Linear Formula CH2CHC(O)NHC(CH3)2CH2C(O)CH3
Formula Weight 169.23
MDL Number MFCD00008788
Synonym N-(1, 1-Dimethyl-3-oxobutyl)acrylamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC352320050

  SDS
thermo fisher scientific
352320050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.23
Physical Form Crystalline Powder or Flakes
Assay ≥98.5%
Quantity 5g
Color White to cream
TSCA TSCA
RTECS Number AS3475000
EINECS Number 220-713-2
Melting Point 53° to 57°C
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Synonym N-(1, 1-Dimethyl-3-oxobutyl)acrylamide
Health Hazard 1 HYGROSCOPIC
Linear Formula CH2CHC(O)NHC(CH3)2CH2C(O)CH3
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Chemical Name or Material Diacetone acrylamide
MDL Number MFCD00008788
CAS 2873-97-4
Name Note 99%
Assay Percent Range 99%
Molecular Formula C9H15NO2
Safety and Handling

HYGROSCOPIC

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.