Please login to your online account to display your discounted pricing

Diacetone acrylamide, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  352320050

 View more versions of this product

Catalog No. AC352320050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Diacetone acrylamide
Quantity 5g
Name Note 99%
CAS 2873-97-4
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number AS3475000
EINECS Number 220-713-2
TSCA TSCA
Physical Form Crystalline Powder or Flakes
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H15NO2
Linear Formula CH2CHC(O)NHC(CH3)2CH2C(O)CH3
Formula Weight 169.23
Infrared Spectrum Authentic
MDL Number MFCD00008788
Synonym N-(1, 1-Dimethyl-3-oxobutyl)acrylamide
Packaging Glass bottle
Color White to cream
Melting Point 53° to 57°C

Specifications

Chemical Name or Material Diacetone acrylamide
Name Note 99%
CAS 2873-97-4
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H15NO2
Linear Formula CH2CHC(O)NHC(CH3)2CH2C(O)CH3
Formula Weight 169.23
MDL Number MFCD00008788
Synonym N-(1, 1-Dimethyl-3-oxobutyl)acrylamide
Accessories

HYGROSCOPIC

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents