Learn More
Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin
CAS: 25395-31-7 | C7H12O5
Supplier: Thermo Scientific Chemicals L127080B
Description
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
| Diacetin | |
| 25395-31-7 | |
| 250°C to 280°C (decomposition) | |
| remainder triacetin and monoacetin | |
| 1.444 | |
| 1000 g | |
| Hygroscopic | |
| glyceryl diacetate, 1,2-diacetin, glycerol diacetate, 1,2,3-propanetriol, diacetate, 3-hydroxypropane-1,2-diyl diacetate, glycerol 1,2-diacetate, 1,2-diacylglycerol, 2,3-diacetin, 1,2-diacetylglycerol, acetin, di | |
| 176.17 | |
| Technical |
| mixed isomers | |
| 1.19 | |
| 141°C (285°F) | |
| C7H12O5 | |
| MFCD00008717 | |
| 1706903 | |
| 14,2961 | |
| Fully miscible in water. | |
| ∼50% |
Chemical Identifiers
| 25395-31-7 | |
| C7H12O5 | |
| MFCD00008717 | |
| glyceryl diacetate, 1,2-diacetin, glycerol diacetate, 1,2,3-propanetriol, diacetate, 3-hydroxypropane-1,2-diyl diacetate, glycerol 1,2-diacetate, 1,2-diacylglycerol, 2,3-diacetin, 1,2-diacetylglycerol, acetin, di |
Safety and Handling
EINECSNumber : 246-941-2
RTECSNumber : AK3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only