Please login to your online account to display your discounted pricing

Alfa Aesar™ Diacetatobis(triphenylphosphine)palladium(II), Pd 14.2%

Chemical Identifiers

Chemical Name or Material Diacetatobis(triphenylphosphine)palladium(II)
CAS 14588-08-0
Linear Formula Pd[P(C6H5)3]2·2CH3CO2
Formula Weight 749.09
MDL Number MFCD00010013
Synonym Bis(triphenylphosphine)palladium(II) acetate; Pd(OAc)2(PPh3)2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6185303

 
alfa aesar™
H6185303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6185309

 
alfa aesar™
H6185309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 749.09
Chemical Name or Material Diacetatobis(triphenylphosphine)palladium(II)
Physical Form Solid
Linear Formula Pd[P(C6H5)3]2·2CH3CO2
MDL Number MFCD00010013
CAS 14588-08-0
Synonym Bis(triphenylphosphine)palladium(II) acetate; Pd(OAc)2(PPh3)2
Melting Point 136°C
Concentration Pd 14.2%
Quantity 1g