Please login to your online account to display your discounted pricing

Alfa Aesar™ Diacetato[1,3-bis(diphenylphosphino)propane]palladium(II), Pd 16.7%

Chemical Identifiers

Chemical Name or Material Diacetato[1,3-bis(diphenylphosphino)propane]palladium(II)
CAS 149796-59-8
Formula Weight 636.96
MDL Number MFCD14155705
Synonym Pd(OAc)2(dppp)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4637003

 
alfa aesar™
4637003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 636.96
Chemical Name or Material Diacetato[1,3-bis(diphenylphosphino)propane]palladium(II)
Physical Form Crystalline Solid
MDL Number MFCD14155705
CAS 149796-59-8
Synonym Pd(OAc)2(dppp)
Color Cream
Concentration Pd 16.7%
Sensitivity Moisture Sensitive
Quantity 1g