Please login to your online account to display your discounted pricing

Di-tert-butyltin dichloride, 98%, ACROS Organics™

$62.00 - $62.00

Chemical Identifiers

Chemical Name or Material Di-tert-butyltin dichloride
Assay Percent Range 98%
Molecular Formula C8H18Cl2Sn
Linear Formula [(CH3)3C]2SnCl2
Formula Weight 303.83
MDL Number MFCD00009854
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC316740010

  SDS
thermo fisher scientific
316740010
Glass bottle 1g Each for $62.00
Description & Specifications

Specifications

Formula Weight 303.83
Physical Form Crystals or Crystalline Powder
Quantity 1g
Color White
Melting Point 39° to 43°C
Linear Formula [(CH3)3C]2SnCl2
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H18Cl2Sn
Chemical Name or Material Di-tert-butyltin dichloride
CAS 19429-30-2
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00009854
Elemental Analysis 97.5% min.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.