Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-tert-butylphosphinoferrocene, 95%

$195.41 - $195.41

Chemical Identifiers

Chemical Name or Material Di-tert-butylphosphinoferrocene
CAS 223655-16-1
Molecular Formula C18H27FeP
Formula Weight 330.23g/mol
MDL Number MFCD06740718
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2918303

 
alfa aesar™
H29183-03
1g Each for $195.41
Description & Specifications

Specifications

Melting Point 91° to 94°C
Quantity 1g
Chemical Name or Material Di-tert-butylphosphinoferrocene
CAS 223655-16-1
Assay 95%
Molecular Formula C18H27FeP
Formula Weight 330.23g/mol
MDL Number MFCD06740718