Please login to your online account to display your discounted pricing

Di-tert-butylphosphine, 0.5M (10 wt.%) solution in hexanes , ACROS Organics™

$148.42 - $148.42

Chemical Identifiers

Chemical Name or Material Di-tert-butylphosphine
CAS 819-19-2
CAS 110-54-3
Molecular Formula C8H19P
Linear Formula [(CH3)3C)2PH
Formula Weight 146.21
MDL Number MFCD00134241
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383020100

  SDS
thermo fisher scientific
AC383020100
Glass bottle 10mL Each for $148.42

AC383020500

  SDS
thermo fisher scientific
AC383020500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 146.21
Quantity 10mL
Color Clear colorless
Density 0.711g/mL
CAS 110-54-3
Chemical Name or Material Di-tert-butylphosphine
Packaging Glass bottle
Molecular Formula C8H19P
Linear Formula [(CH3)3C)2PH
MDL Number MFCD00134241
CAS 819-19-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
NMR Complies
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Suspected of dama
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Suspected of dama