Please login to your online account to display your discounted pricing

Alfa Aesar™ Di-tert-butylphenylphosphonium tetrafluoroborate, 99%

$48.66 - $113.76

Chemical Identifiers

Chemical Name or Material Di-tert-butylphenylphosphonium tetrafluoroborate
CAS 612088-55-8
Molecular Formula [(CH3)3C]2PH(C6H5)·BF4
Formula Weight 310.12
MDL Number MFCD08704553
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2690403

  SDS
alfa aesar™
H2690403
1g Each for $113.76

AAH26904MD

  SDS
alfa aesar™
H26904MD
250mg Each for $48.66
Description & Specifications

Specifications

Formula Weight 310.12
Chemical Name or Material Di-tert-butylphenylphosphonium tetrafluoroborate
Assay 99%
Molecular Formula [(CH3)3C]2PH(C6H5)·BF4
MDL Number MFCD08704553
CAS 612088-55-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.