Please login to your online account to display your discounted pricing

Di-tert-Butylchlorosilane, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC432900250

 View more versions of this product

Catalog No. AC432900250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94%
Formula Weight 178.78
Quantity 25g
Color Clear colorless
Density 0.8840g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C8H19ClSi
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Contact with water liberates toxic gas.
Chemical Name or Material Di-tert-Butylchlorosilane
CAS 56310-18-0
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Handle under inert gas. Protect from moisture.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Di-tert-Butylchlorosilane
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H19ClSi
Formula Weight 178.78
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Handle under inert gas. Protect from moisture.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases.
Contact with water liberates toxic gas.